Order Entry
Northern Ireland
ContactUsLinkComponent
Benzophenone imine
Benzophenone imine
Catalog # MOLEM28942128
Supplier:  Molekula
CAS Number:  
Benzophenone imine
Catalog # MOLEM28942128
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :181.24 g/mol
CAS Number:1013-88-3
Formula:C₁₃H₁₁N
MDL Number:MFCD00001760