Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chloro-4,6-dimethoxy-1,3,5-triazine
2-Chloro-4,6-dimethoxy-1,3,5-triazine
Catalog # MOLE10809468-25G
Supplier:  Molekula
CAS Number:  
2-Chloro-4,6-dimethoxy-1,3,5-triazine
Catalog # MOLE10809468-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :175.57 g/mol
EINECS:221-541-0
CAS Number:3140-73-6
Formula:C₅H₆ClN₃O₂
MDL Number:MFCD00075607