Methyl-3-methoxy-2-nitrobenzoate ≥98%
Catalog # L08886.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:211.17 g/mol
Melting Pt:142…144 °C
Storage Temperature:Ambient
CAS Number:5307-17-5
Formula:C₉H₉NO₅
MDL Number:MFCD00051968