
Zinc(II) acetylacetonate monohydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14363-15-6
:MFCD00000035
Formula:C₁₀H₁₄O₄Zn·1H₂O
:281.62 g/mol
Melting Pt:136…138 °C (dec.)
Density:1,6
EINECS:237-860-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together