Order Entry
Northern Ireland
ContactUsLinkComponent
2-Ethylhexyl methacrylate 99% stabilised
Irritant
2-Ethylhexyl methacrylate 99% stabilised
Certificates
undefined
Irritant
2-Ethylhexyl methacrylate 99% stabilised

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4370 to 1.4390 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA
  • Stabilizer:
    40 to 60 ppm MEHQ

Specification Test Results

Related Information

Frequently Bought Together