4-Methoxybenzaldehyde dimethyl acetal 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2186-92-7
:
Formula:C₁₀H₁₄O₃
MDL Number:MFCD00036507
Boiling Pt:235 °C (760 mmHg)
Density:1,068 g/cm³
Flash Pt:>110 °C
:182.22 g/mol
Storage Temperature:Refrigerator
EINECS:218-577-4
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5035 to 1.5075 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together