

4-Fluorophenylhydrazinium chloride 97%
:823-85-8
:
:C₆H₈ClFN₂
:MFCD00012942
:162.59 g/mol
:275…277 °C
:Ambient
:212-521-2
:2811
:6.1,III
:GHS07
:GHS08
- Appearance:White to brown Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
- GC:≥96 %
- NMR (1H-NMR):Consistent with structure
Frequently Bought Together