

1-Bromo-4-fluorobenzene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:460-00-4
:
Formula:C₆H₄BrF
MDL Number:MFCD00000342
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–8 °C
Density:1,593 g/cm³
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:207-300-2
ADR:3,III
:175 g/mol
UN:1992
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5260 to 1.5280 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together