

2,4,4-Trimethyl-1-pentene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-39-1
:
Formula:C₈H₁₆
MDL Number:MFCD00008855
:112.22 g/mol
Boiling Pt:101…102 °C (760 mmHg)
Melting Pt:–101 °C
Density:0,7 g/cm³
Flash Pt:–6 °C
Storage Temperature:Ambient
EINECS:203-486-4
UN:2050
ADR:3,II
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4090 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:+25° to +30° (20°C, 589 nm) (c=1 in water)
Specification Test Results
Frequently Bought Together