

2,3-Dimethyl-2-butene ≥97%
Catalog # A13201.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:84.16 g/mol
Boiling Pt:72…73 °C
Melting Pt:–75 °C
Density:0,705
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:209-263-8
UN:3295
ADR:3,II
:GHS02
:GHS08
CAS Number:563-79-1
Formula:C₆H₁₂
MDL Number:MFCD00008897