



3-Chloro-2-methylpropene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:563-47-3
:
Formula:C₄H₇Cl
MDL Number:MFCD00000953
:90.55 g/mol
Merck Index:13,02167
Boiling Pt:72 °C (1013 hPa)
Melting Pt:–80 °C
Density:0,93 g/cm³ (20 °C)
UN:2554
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:209-251-2
:GHS02
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.5 g/l (20 °C)
- Melting Point:-80 °C
- Molar Mass:90.55 g/mol
- Boiling Point:72 - 74 °C
- Vapor Pressure:140 hPa (20 °C)
- Flash Point:-18 °C
- Refractive Index:1.4272 (20 °C, 589 nm)
- Explosion Limit:2.2 - 10.4 %(V)
- Density:0.93 g/cm3 (20 °C)
- Ignition Point:540 °C
Specification Test Results
Frequently Bought Together