

cis-4-Octene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7642-15-1
:
Formula:C₈H₁₆
MDL Number:MFCD00067484
:112.22 g/mol
Boiling Pt:123 °C (1013 hPa)
Melting Pt:–118,7 °C
Density:0,7212 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-578-4
UN:3295
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together