Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
1-Dodecene for synthesis, Sigma-Aldrich®
 :  Merck
 
 
1-Dodecene for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -35 °C
  • Molar Mass:
    168.32 g/mol
  • Boiling Point:
    213 - 216 °C (1013 hPa)
  • Vapor Pressure:
    0.23 hPa (20 °C)
  • Flash Point:
    77 °C
  • Refractive Index:
    1.4294 (20 °C, 589 nm)
  • Explosion Limit:
    0.6 - 5.4 %(V)
  • Density:
    0.76 g/cm3 (20 °C)
  • Ignition Point:
    226 °C
  • Viscosity kinematic:
    1.71 mm2/s (20 °C)

 

Frequently Bought Together