
1,1-Bis(methylthio)-2-nitroethylene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13623-94-4
:
Formula:C₄H₇NO₂S₂
MDL Number:MFCD00010443
:165.24 g/mol
Melting Pt:124…126 °C
Storage Temperature:Ambient
EINECS:237-108-4
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Melting point:124 to 127 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together