1H,1H,2H-Perfluoro-1-dodecene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30389-25-4
:
Formula:C₁₂H₃F₂₁
MDL Number:MFCD00042346
:546.12 g/mol
Boiling Pt:71…72 °C (14 mmHg)
Density:1,711
Storage Temperature:Ambient
EINECS:250-173-3
Frequently Bought Together