ContactUsLinkComponent
Filtration capsules, Sartoclean® CA
Filtration capsules, Sartoclean® CA
Supplier:  Sartorius
Certificates
undefined
Filtration capsules, Sartoclean® CA
Supplier:  Sartorius
Certificates
Specifications
  • Catalog No:
  • SARP5625304E9-SS-A
  • SARP5625304E9-OO-A
  • Inlet/outlet:
  • SS (1 1/2" sanitary flange/1 1/2" sanitary flange)
  • OO (1/2'' single stepped hose barb/1/2'' single stepped hose barb)
  • Ø×L:
  • 108,5×197 mm
  • 108,5×210 mm
  • Max. pressure:
  • 5 bar
  • 5 bar