Methyl phenylacetate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:101-41-7
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008453
:150.18 g/mol
Merck Index:14,7268
Boiling Pt:218…220 °C
Melting Pt:49…50 °C
Density:1,067
Flash Pt:85 °C (185 °F)
Storage Temperature:Ambient
EINECS:202-940-9
Related Information
Frequently Bought Together