2,3-Dimethylpentane ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 565-59-3 Molecular Formula: C7H16 Formula Weight: 100.21 Flash Point: -6°(21°F) Physical Form: Liquid Density: 0.690 Refractive Index: 1.3920 Boiling Point: 89-90° UN No.: 1206 MDL No.: MFCD00009337
- CAS Number:565-59-3
- Formula:C₇H₁₆
- MDL Number:MFCD00009337
- Molecular Weight:100.2 g/mol
- Boiling Pt:89…90 °C
- Melting Pt:–135 °C
- Density:0,690
- Flash Pt:–6 °C (21 °F)
- Storage Temperature:Ambient
- EINECS:209-280-0
- UN:1206
- ADR:3,II
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 565-59-3 Molecular Formula: C7H16 Formula Weight: 100.21 Flash Point: -6°(21°F) Physical Form: Liquid Density: 0.690 Refractive Index: 1.3920 Boiling Point: 89-90° UN No.: 1206 MDL No.: MFCD00009337
- CAS Number:565-59-3
- Formula:C₇H₁₆
- MDL Number:MFCD00009337
- Molecular Weight:100.2 g/mol
- Boiling Pt:89…90 °C
- Melting Pt:–135 °C
- Density:0,690
- Flash Pt:–6 °C (21 °F)
- Storage Temperature:Ambient
- EINECS:209-280-0
- UN:1206
- ADR:3,II
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



