Order Entry
Northern Ireland
ContactUsLinkComponent
Stopcocks, needle-valve, straight
Stopcocks, needle-valve, straight
  227-3310
undefined
Stopcocks, needle-valve, straight
  227-3310

 

  • Hole Ø:
    0 - 2,5 mm
  • Pk:
    1

 

 

Needle-valve stopcocks are made of DURAN® borosilicate glass, with PTFE spindle.

  • Screw cap and retention device made of PP
  • Temperature resistance from −20 to +150 °C
  • Excellent chemical resistance

These stopcocks are ideal for exact dispensing and therefore ideal for glass equipment.