2,2,4-Trimethylpentane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
Boiling Pt:98…99 °C
Melting Pt:–107 °C
Density:0.692
Molecular Weight:114.23 g/mol
Flash Pt:–7 °C (19 °F)
UN:1262
ADR:3 II
Merck Index:13,05212
EINECS:208-759-1
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together