

5-Methyl-2-hexanone for synthesis, Sigma-Aldrich®
: Merck
:110-12-3
:
:C₇H₁₄O
:MFCD00008950
:114.19 g/mol
:144 °C (1013 hPa)
:–74 °C
:0,81 g/cm³ (20 °C)
:43 °C
:2302
:3,III
:M-Clarity™ quality level = MQ200
:203-737-8
:GHS02
:GHS07
:Ambient
- Solubility:5.4 g/l (20 °C)
- Melting Point:-74 °C
- Molar Mass:114.19 g/mol
- Boiling Point:144 °C (1013 hPa)
- Vapor Pressure:5.3 hPa (20 °C)
- Flash Point:43 °C
- Refractive Index:1.4078 (20 °C, 589 nm)
- Explosion Limit:1.4 %(V)
- Density:0.81 g/cm3 (20 °C)
- Ignition Point:455 °C
Frequently Bought Together