Isobutyraldehyde 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-84-2
:
Formula:C₄H₈O
MDL Number:MFCD00006980
:72.11 g/mol
Merck Index:13,05171
Boiling Pt:63 °C (760 mmHg)
Melting Pt:–65 °C
Density:0,79 g/cm³
Flash Pt:–24 °C
Storage Temperature:Refrigerator
EINECS:201-149-6
UN:2045
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3710 to 1.3745 (20°C, 589 nm)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Free acid:≤1.0 % (Isobutyric acid)
Specification Test Results
Frequently Bought Together