

Neopentyl bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:630-17-1
:
Formula:C₅H₁₁Br
MDL Number:MFCD00000209
:151.05 g/mol
Boiling Pt:104…106 °C
Density:1,195
Flash Pt:6 °C (42 °F)
Storage Temperature:Ambient
EINECS:211-132-5
UN:1993
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together