ContactUsLinkComponent
n-Pentane 99+%, ECD tested for pesticide analysis for residue analysis
Irritant
Health
Environment
Flammable
n-Pentane 99+%, ECD tested for pesticide analysis for residue analysis
Certificates
undefined
Irritant
Health
Environment
Flammable
n-Pentane 99+%, ECD tested for pesticide analysis for residue analysis
Specification Test Results
  • Appearance:
    Clear liquid
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤5 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity (CH3COOH):
    ≤0.003 %