

Methyl-3-(trimethylsilyl)propiolate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:42201-71-8
:
Formula:C₇H₁₂O₂Si
MDL Number:MFCD04039783
:156.26 g/mol
Boiling Pt:71…75 °C (11 mmHg)
UN:3272
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together