



n-Nonane 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-84-2
:
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574
Boiling Pt:151 °C (760 mmHg)
Melting Pt:–53 °C
Density:0,718 g/cm³
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:203-913-4
ADR:3,III
:128.26 g/mol
UN:1920
:GHS02
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together