Hexane (mixture of isomers) ≥98.5% ACS
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD02179311
:86.18 g/mol
:13,04712
:65…70 °C
:–95 °C
:0,67
:<–23 °C (–9 °F)
:295-570-2
:1208
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
Frequently Bought Together