n-Hexacosane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:630-01-3
:
Formula:H₃C(CH₂)₂₄CH₃
MDL Number:MFCD00009354
:366.71 g/mol
Boiling Pt:412,2 °C (1013 hPa)
Melting Pt:56…58 °C
Density:0,804 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-124-1
Frequently Bought Together