n-Heptane 99+%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: high purity solvents
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
Boiling Pt:98.4
Melting Pt:–91 °C
Density:0.683 g/cm³
Molecular Weight:100.20 g/mol
Flash Pt:–4 °C
UN:1206
ADR:3 II
Merck Index:13,04679
EINECS:205-563-8
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- GC/ECD (as lindane):≤5 ng/l
- Residue after evaporation:≤0.0003 %
- Acidity (CH3COOH):≤0.002 %
Specification Test Results
Frequently Bought Together