Supelco® Organic Reference Standard, Heneicosane
: Merck
- CAS No.:629-94-7
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00009346
- M-Clarity quality level:MQ100
- Melting point:39 - 41 °C (lit.); 39 - 42 °C
- Formula:CH₃(CH₂)₁₉CH₃
- Molecular weight:296,57
- Flash point:113 °C - Closed cup
- Boiling point:100 °C/2 mmHg (lit.); 356,1 °C/760 mmHg
Frequently Bought Together