
1-Octadecanethiol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2885-00-9
:
Formula:C₁₈H₃₈S
MDL Number:MFCD00004886
:286.57 g/mol
Boiling Pt:180 °C (5 mmHg)
Melting Pt:30…33 °C
Density:0,847
Flash Pt:185 °C (365 °F)
Storage Temperature:Ambient
EINECS:220-744-1
UN:3335
ADR:9,
:GHS07
Related Information
Frequently Bought Together