Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
1,8-Octanediol 98%

1,8-Octanediol 98%

Irritant
1,8-Octanediol 98%
Certificates
1,8-Octanediol 98%

2 Options Available Below

Product Details & Documents
Synonyms:  1,8-Dihydroxy octane
  • CAS Number:629-41-4
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00002989
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:278,8 °C (760 mmHg)
  • Melting Pt:60…62 °C
  • Density:0,941 g/cm³ (20 °C)
  • Flash Pt:128 °C
  • Storage Temperature:Ambient
  • EINECS:211-090-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to white Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 63 °C
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £55.40
    £55.40
    1 * 25 g
       £55.40

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £161.00
    £161.00
    1 * 100 g
       £161.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.