Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Methanol

Methanol

Plastic can, 5 L
Catalog # 40296-5L
Flammable
Toxic
Health
Supplier:   Honeywell Chemicals
Methanol
Catalog # 40296-5L
CAS Number:  67-56-1
Methanol
Methanol

Plastic can, 5 L

CAS Number:  67-56-1
Price
£286.00
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64,6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0,7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44

Documents

Specifications

Specifications

  • Pack type:
    Plastic can
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.