


1-Propanol, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:71-23-8
:
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941
:60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0,8053 g/cm³ (20 °C)
EINECS:200-746-9
UN:1274
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:15 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together