1-Octanol ≥98%, purified
Catalog # 20850.296
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 98 %
- Boiling point:194 - 196 °C
- Density (20/4):0.824 - 0.827
Specification Test Results
About this item
:
:130.23 g/mol
Merck Index:13,06784
Boiling Pt:195 °C (1013 hPa)
Melting Pt:–16 °C
Density:0,8246 g/cm³ (20 °C)
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:203-917-6
:GHS07
CAS Number:111-87-5
Formula:H₃C(CH₂)₆CH₂OH
MDL Number:MFCD00002988