Order Entry
Northern Ireland
ContactUsLinkComponent
Arachidyl alcohol for synthesis
Arachidyl alcohol for synthesis
Catalog # 8.22051.0100
Supplier:  Merck
CAS Number:  
Arachidyl alcohol for synthesis
Catalog # 8.22051.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    65 °C
  • Molar Mass:
    298.55 g/mol
  • Boiling Point:
    220 - 225 °C (4 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    195 °C
  • Ignition Point:
    245 °C
  • Synonyms:
    Arachyl alcohol
  • Hill Formula:
    C20H42O
  • Chemical Formula:
    CH3(CH2)19OH

Specification Test Results

About this item

 :
 :298.55 g/mol
Boiling Pt:309 °C (1013 hPa)
Melting Pt:71,5 °C
Density:0,88 g/cm³ (21 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-119-4
CAS Number:629-96-9
Formula:H₃C(CH₂)₁₈CH₂OH
MDL Number:MFCD00002938