

Diethyl ether 99+% inhibitor-free for spectrophotometry Inhibitor-free
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:14,3806
Boiling Pt:34,6 °C
Melting Pt:–116 °C
Density:0,713
Flash Pt:–45 °C (–49 °F)
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
:GHS02
:GHS07
Related Information
Frequently Bought Together