Order Entry
Northern Ireland
ContactUsLinkComponent
Octadecylamine for synthesis, Sigma-Aldrich®
Corrosive
Health
Environment
Octadecylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Health
Environment
Octadecylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    50 - 52 °C
  • Molar Mass:
    269.51 g/mol
  • Boiling Point:
    349 °C
  • Vapor Pressure:
     - 100 hPa (20 °C)
  • Flash Point:
    110 °C
  • Density:
    0.86 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together