Order Entry
Northern Ireland
ContactUsLinkComponent
Isopropyl isocyanate ≥98%
Flammable
Toxic
Health
Irritant
Isopropyl isocyanate ≥98%
Catalog # L16105.03
CAS Number:  
Flammable
Toxic
Health
Irritant
Isopropyl isocyanate ≥98%
Catalog # L16105.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :85.11 g/mol
Boiling Pt:74…75 °C
Density:0,868
Flash Pt:–3 °C (26 °F)
Storage Temperature:Refrigerator
EINECS:217-276-5
UN:2483
ADR:6.1,I
 :GHS02
 :GHS06
 :GHS08
 :GHS07
CAS Number:1795-48-8
Formula:C₄H₇NO
MDL Number:MFCD00002037