Order Entry
Northern Ireland
ContactUsLinkComponent
Octyl isocyanate ≥97%
Toxic
Health
Irritant
Octyl isocyanate ≥97%
Catalog # L18746.06
CAS Number:  
Toxic
Health
Irritant
Octyl isocyanate ≥97%
Catalog # L18746.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :155.24 g/mol
Boiling Pt:200…204 °C
Density:0,880
Flash Pt:71 °C (159 °F)
Storage Temperature:Ambient
EINECS:221-598-7
UN:2206
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS07
CAS Number:3158-26-7
Formula:C₉H₁₇NO
MDL Number:MFCD00013887