Order Entry
Northern Ireland
ContactUsLinkComponent
2-Iodopropane 98+% stabilised
Irritant
Flammable
2-Iodopropane 98+% stabilised
Certificates
undefined
Irritant
Flammable
2-Iodopropane 98+% stabilised
Certificates

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4972 to 1.4992 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Stabilizer:
    copper
  • Elemental Analysis:
    6.15 to 6.94 % (N)

Specification Test Results

Related Information

Frequently Bought Together