
1,2-Diiodoethane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:624-73-7
:
Formula:C₂H₄I₂
MDL Number:MFCD00001093
:281.86 g/mol
Boiling Pt:206 °C (1013 hPa)
Melting Pt:80…83 °C
Density:2,13 g/cm³
Storage Temperature:Refrigerator
EINECS:210-859-5
:GHS07
Specification Test Results
- Appearance:Brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:80 to 83 °C
- GC:≥98.5 %
- Clarity of solution:Passes test
- Substance darkened by H2SO4:Passes test
- Chloride (Cl):≤20 ppm
- Nitrate (NO3):≤20 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤200 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.01 %
Specification Test Results
Frequently Bought Together