

2-Chloroethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-07-3
:
Formula:C₂H₅ClO
MDL Number:MFCD00002829
:80.51 g/mol
Merck Index:13,03827
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–67,5 °C
Density:1,204 g/cm³ (20 °C)
EINECS:203-459-7
UN:1135
ADR:6.1 (3),I
:M-Clarity™ quality level = MQ200
Flash Pt:28 °C
:GHS02
:GHS06
Storage Temperature:Ambient
Frequently Bought Together