
5-Chloro-2-pentanone
Catalog # MOLE49200869-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:120.58 g/mol
Boiling Pt:71 °C (1013 hPa)
Melting Pt:–48 °C
Density:1,057 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:227-565-8
UN:1224
ADR:3,III
:GHS07
CAS Number:5891-21-4
Formula:C₅H₉ClO
MDL Number:MFCD00001008