Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chloropropane for synthesis, Sigma-Aldrich®
Flammable
Irritant
2-Chloropropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
2-Chloropropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    3.1 g/l (20 °C)
  • Melting Point:
    -117 °C
  • Molar Mass:
    78.54 g/mol
  • Boiling Point:
    36 °C (1013 hPa)
  • Vapor Pressure:
    580 hPa (20 °C)
  • Flash Point:
    -36 °C
  • Refractive Index:
    1.3778 (20 °C, 589 nm)
  • Explosion Limit:
    2.8 - 10.7 %(V)
  • Density:
    0.86 g/cm3 (20 °C)
  • Ignition Point:
    590 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together