

1-Chlorodecane 97-99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1002-69-3
:
Formula:C₁₀H₂₁Cl
MDL Number:MFCD00000956
:176.73 g/mol
Boiling Pt:223 °C (760 mmHg)
Melting Pt:–34 °C
Density:0,86 g/cm³
Flash Pt:83 °C
Storage Temperature:Ambient
EINECS:213-691-0
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4362 to 1.4382 (20°C, 589 nm)
- GC:≥97 %
- Residue after evaporation:≤0.003 %
- Carbonyl (as HCHO):≤0.1 %
- Acids and esters:≤0.2 % (as Amyl acetate)
- Titrable acid:≤0.002 meq/g
Specification Test Results
Frequently Bought Together