1-Chlorobutane for synthesis, Sigma-Aldrich®
: Merck
:109-69-3
:
:CH₃(CH₂)₃Cl
:MFCD00001009
:92.57 g/mol
:13,01559
:78,4 °C (1013 hPa)
:–123 °C
:0,8862 g/cm³ (20 °C)
:Ambient
:203-696-6
:1127
:3,II
:M-Clarity™ quality level = MQ200
:–17 °C
:GHS02
- Solubility:0.5 g/l (20 °C)
- Melting Point:-123 °C
- Molar Mass:92.57 g/mol
- Boiling Point:79 °C (1013 hPa)
- Vapor Pressure:110 hPa (20 °C)
- Flash Point:-17 °C
- Explosion Limit:1.8 - 10.1 %(V)
- Density:0.886 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:280 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together