
1,4-Dichlorobutane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-56-5
:
Formula:ClH₂C(CH₂)₂CH₂Cl
MDL Number:MFCD00001011
:127.01 g/mol
Boiling Pt:155 °C
Melting Pt:–38 °C
Density:1,139
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
EINECS:203-778-1
UN:1993
ADR:3,III
:GHS02
Related Information
Frequently Bought Together