Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
1,4-Dichlorobutane 99%

1,4-Dichlorobutane 99%

Flammable
1,4-Dichlorobutane 99%
Certificates
1,4-Dichlorobutane 99%

3 Options Available Below

Product Details & Documents
  • CAS Number:110-56-5
  • Formula:ClH₂C(CH₂)₂CH₂Cl
  • MDL Number:MFCD00001011
  • Molecular Weight:127.01 g/mol
  • Boiling Pt:161…163 °C (760 mmHg)
  • Melting Pt:–38 °C
  • Density:1,16 g/cm³
  • Flash Pt:40 °C
  • Storage Temperature:Ambient
  • EINECS:203-778-1
  • UN:1993
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4550 (20°C, 589 nm)
  • Water:
    ≤0.05 %
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - £55.90
    £55.90
    1 * 100 mL
       £55.90

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - £161.00
    £161.00
    1 * 500 mL
       £161.00

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - £259.00
    £259.00
    1 * 1 L
       £259.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.