1,2-Dichloroethane
Catalog # PROOCHE19
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,5 mL
- Seq:2095
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ethylene chloride, Ethylene dichloride, Dichloroethane
:
:98.96 g/mol
Flash Pt:13
EINECS:203-458-1
UN:1184
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:107-06-2
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963
Boiling Pt:83.5
Melting Pt:–35.5 °C
Density:1.254 g/cm³ (20 °C)
ADR:3 II
Merck Index:13,03831